Anti-PGR Mouse Monoclonal Antibody [clone: PR500]
Lieferant: ProSci Inc.
33-189
PRSI33-189EA 773 EUR
PRSI33-189
Anti-PGR Mouse Monoclonal Antibody [clone: PR500]
Antikörper
This antibody is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of progesterone receptor has been suggested to reflect a intact estrogen regulatory machinery and therefore predict better clinical response to endocrine therapy than ER alone.
Immunohistochemistry: 0,5 - 1 µg/ml
The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be
More Product Information
Neuer Artikel bei VWR!