Anti-CD176 Mouse Monoclonal Antibody [clone: SPM320]

Lieferant: ProSci Inc.
33-533
PRSI33-533EA 773 EUR
PRSI33-533
Anti-CD176 Mouse Monoclonal Antibody [clone: SPM320]
Antikörper
Recognises a disaccharide epitope, Gal1-3GalNAc, of Thomsen-Friedenreich (TF) antigen. It is specific for both anomeric forms of the disaccharide (TF and TF, including related structures on the glycolipid) and shows no cross-reactivity with sialylated glycophorin. The Thomsen-Friedenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered as a pan-carcinoma marker. During metastasis, the ability of malignant cells to form multicellular aggregates via homotypic or heterotypic aggregation and their adhesion to the endothelium are critical. The tumor-associated carbohydrate Thomsen-Friedenreich antigen (Gal-GalNAc) is involved in tumor cell adhesion and tissue invasion. It also causes an immune response, and overexpression of the antigen causes cancer cells to be more sensitive to natural killer cell lysis. The Thomsen-Friedenreich antigen is suppressed in normal healthy cells and represents one of the few chemically well-defined antigens associated with tumor malignancy. The presence of the Thomsen-Friedenreich antigen on the surface of cancer cells may result from a divergence from the normal pathway for O-linked glycosylation in these cells, most likely caused by inappropriate localization of the enzymes involved in synthesis of the disaccharide.

Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 3-5 µg/ml for 30 min at RT (1) Optimal dilution of the CD176 antibody should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 min.


Type: Primary
Antigen: CD176 Antibody [SPM320]
Clonality: Monoclonal
Clone: SPM320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR