Anti-IGLV1-51, cycH Mouse Monoclonal Antibody [clone: LLC/1738]

Lieferant: ProSci Inc.
33-633
PRSI33-633EA 773 EUR
PRSI33-633
Anti-IGLV1-51, cycH Mouse Monoclonal Antibody [clone: LLC/1738]
Antikörper
This mAb is specific to lambda light chain of immunoglobulin and shows no cross-reaction with kappa light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT The optimal dilution of the Lambda Light Chain antibody for each application should be determined by the researcher.

Type: Primary
Antigen: IGLV1-51, cycH
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR