Anti-MALT1 Mouse Monoclonal Antibody [clone: 4NMT-3]

Lieferant: ProSci Inc.
33-934
PRSI33-934EA 773 EUR
PRSI33-934
Anti-MALT1 Mouse Monoclonal Antibody [clone: 4NMT-3]
Antikörper
Mucosa associated lymphoid tissue lymphoma translocation gene 1 is found in extranodal low-grade B cell lymphomas. It encodes two Ig-like C2-type domains and fuses with an API2 gene, which is highly expressed in adult lymphoid tissue. The translocation of the MALT1 gene, which maps to human chromosome 18q21, and the apoptosis-inhibiting API2 gene results in an increased development of MALT lymphomas and apoptosis inhibition.

IF: 1 - 2 µg/ml WB: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) Titering of the MALT1 antibody may be required for optimal performance.

1. FFPE testing requires sections to be boiled in pH6 10 mM citrate buffer for 10 - 20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: MALT1
Clonality: Monoclonal
Clone: 4NMT-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR