Anti-PIKE Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
3321
PRSI3321EA 435 EUR
PRSI3321
Anti-PIKE Rabbit Polyclonal Antibody
Antikörper
PIKE Antibody: Phosphoinositide 3 kinase enhancer (PIKE) is a recently identified nuclear GTPase that interacts with nuclear phosphoinositide 3-kinase (PI3 kinase) to stimulate its lipid kinase activity. PIKE exists in multiple isoforms; a shorter C-terminal isoform (PIKE-A) has also been identified as centaurin gamma 1. The longest isoform (PIKE-L) has been shown to bind to the adaptor protein Homer and thereby link to metabotropic glutamate receptors, leading to activation of PI3 kinase activity and prevention of neuronal apoptosis. Overexpression of PIKE-A enhances Akt activity and promotes cancer cell invasion, whereas decreased expression of PIKE-A via dominant negative expression of PIKE-A or PIKE-A knockdown inhibits these processes. In many human cancers, expression of PIKE-A is enhanced, leading to increased Akt activity and preventing apoptosis.

Type:
Antigen: PIKE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR