Anti-Feline Leukemia virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: ProSci Inc.
35-187
PRSI35-187EA 588 EUR
PRSI35-187
Anti-Feline Leukemia virus Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.

Feline Leukemia Virus antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Feline Leukemia virus
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR