Anti-Gram positive bacteria Mouse Monoclonal Antibody

Lieferant: ProSci Inc.
35-578
PRSI35-578EA 588 EUR
PRSI35-578
Anti-Gram positive bacteria Mouse Monoclonal Antibody
Antikörper
Gram positive bacteria antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.

Gram positive bacteria antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Gram positive bacteria
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR