Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 488) [clone: HuBR9.1]
Lieferant: ProSci Inc.
36-117
PRSI36-117EA 686 EUR
PRSI36-117
Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 488) [clone: HuBR9.1]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human