Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 647N) [clone: HuBR9.1]
Lieferant: ProSci Inc.
36-118
PRSI36-118EA 686 EUR
PRSI36-118
Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 647N) [clone: HuBR9.1]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human