Anti-TNFRSF10A Mouse Monoclonal Antibody (ATTO 647N) [clone: HS101]

Lieferant: ProSci Inc.
36-127
PRSI36-127EA 734 EUR
PRSI36-127
Anti-TNFRSF10A Mouse Monoclonal Antibody (ATTO 647N) [clone: HS101]
Antikörper
TRAIL-R1 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: HS101
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR