Anti-TNFRSF13C Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Lieferant: ProSci Inc.
36-144
PRSI36-144EA 686 EUR
PRSI36-144
Anti-TNFRSF13C Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: 9B9
Conjugation: ATTO 647N
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse