Anti-COL18A1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
38-194
PRSI38-194EA 509 EUR
PRSI38-194
Anti-COL18A1 Rabbit Polyclonal Antibody
Antikörper
Endostatin, a cleaved product of the C-terminal domain of collagen XVIII, is an endogenous angiogenesis inhibitor. It was originally found in conditioned media from a murine endothelial tumor cell line, hemangioendothelioma. Endostatin inhibits endothelial cell migration (in vivo and in vitro) and induces endothelial cell apoptosis. It inhibits tumor growth and impairs blood vessel maturation in wound healing. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type:
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR