Anti-NGFRAP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
46-876
PRSI46-876EA 488 EUR
PRSI46-876
Anti-NGFRAP1 Goat Polyclonal Antibody
Antikörper
NGFRAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NGFRAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NGFRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR