Anti-HAX1A Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
4711
PRSI4711EA 435 EUR
PRSI4711
Anti-HAX1A Rabbit Polyclonal Antibody
Antikörper
Hax1a Antibody: The HS-1 associated protein X-1 (Hax1) was initially identified in a yeast two-hybrid assay on the basis of its ability to bind to the hemapoietic cell-specific protein 1 (HS-1). Hax1 possesses anti-apoptotic activity and is structurally related to Bcl-2 family members, including the presence of BH1- and BH2-like domains. It has recently been shown to interact with HIV viral protein R (Vpr), a protein required for viral pathogenesis of HIV and linked to T-cell apoptosis through activation of caspases 3 and 9. Other studies indicate that Hax1-mediated processing of HtrA2 (alsoMore Product Information