Anti-PDE8A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
48-315
PRSI48-315EA 694 EUR
PRSI48-315
Anti-PDE8A Rabbit Polyclonal Antibody
Antikörper
Phosphodiesterase 8A, a high affinity, cAMP-specific phosphodiesterase, has been implicated in penile erectile dysfunction. PDE8A activity is inhibited by dipyridamole, but not by rolipram, zaprinast, or IBMX. At least five splice variants and four protein isoforms have been reported.

PDE8A antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:300.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Horse, Human, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR