Anti-PDE8B Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
48-317
PRSI48-317EA 694 EUR
PRSI48-317
Anti-PDE8B Rabbit Polyclonal Antibody
Antikörper
Phosphodiesterase 8B, is a high affinity, cAMP-specific phosphodiesterase. Enhanced PDE8B activity has been associated with the phenotypic expression of Gsalpha mutations in human pituitary tumors. PDE8B is absent in normal pituitary but present in GH-secreting adenomas. PDE8B activity is inhibited by dipyridamole, but it is insensitive to rolipram, zaprinast, or IBMX.

PDE8B antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR