Anti-PDE9A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
48-318
PRSI48-318EA 694 EUR
PRSI48-318
Anti-PDE9A Rabbit Polyclonal Antibody
Antikörper
Phosphodiesterase 9A is a cGMP-specific phosphodiesterase. PDE9A activity is inhibited by zaprinast, but it is insensitive to rolipram, vinpocetine, SKF-94120, dipyridamole, and IBMX. More than 20 alternatively spliced variants have been reported (Rentero et al. 2003). PDE9A has been implicated in penile erectile dysfunction, as well as in functional disturbances in which intraneuronal signal transmission via second messengers are important in the pathophysiology (example, bipolar affective disorder). PDE9A is the only cGMP-specific PDE with significant expression in the forebrain, and as such is likely to function in NO-cGMP signaling.

PDE9A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR