Anti-SELP Mouse Monoclonal Antibody

Lieferant: ProSci Inc.
49-086
PRSI49-086EA 771 EUR
PRSI49-086
Anti-SELP Mouse Monoclonal Antibody
Antikörper
CD62P antibody can be used in immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SELP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR