Anti-ORAI3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
49-890
PRSI49-890EA 694 EUR
PRSI49-890
Anti-ORAI3 Rabbit Polyclonal Antibody
Antikörper
Recognizes human ORAI3, also known as Transmembrane protein 142C (TMEM142C), a 31.5 kDa multi-pass membrane protein belonging to the ORAI family. Antigen stimulation of immune cells triggers Ca2+ entry through Ca2+ release-activated Ca2+ (CRAC) channels. CRAC channels are the main pathway for Ca2+ influx in T-cells and promote the immune response to pathogens by activating the transcription factor NFAT. ORAI3 is one of two mammalian homologs of ORAI1, a four- transmembrane spanning protein that is an essential component of CRAC channels. ORAI3, along with ORAI1 and ORAI2, functions as a Ca2+ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca2+ sensor. Studies indicate that ORAI3 channels undergo a lesser degree of depotentiation than ORAI1 or ORAI2. Is not expected to cross react with either ORAI1 or ORAI2.

ORAI3 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, immunofluorescence starting at 10 μg/mL, and immunoprecipitation.

Affinity Chromatography

PBS, 0.02% sodium azide.

Store ORAI3 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type:
Antigen: ORAI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR