Anti-Rab3a Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
50-187
PRSI50-187EA 686 EUR
PRSI50-187
Anti-Rab3a Rabbit Polyclonal Antibody
Antikörper
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D (Schluter et al., 2004). These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters (Coppola et al., 2002). Specifically, Rab3’s are thought to play a key role in presynaptic vesicle trafficking and priming (Schluter et al., 2006).

Affinity Purified

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Rab3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR