Anti-NAA25 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-116
PRSI55-116EA 620 EUR
PRSI55-116
Anti-NAA25 Rabbit Polyclonal Antibody
Antikörper
This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: NAA25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR