Anti-BCO1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-200
PRSI55-200EA 620 EUR
PRSI55-200
Anti-BCO1 Rabbit Polyclonal Antibody
Antikörper
Vitamin A metabolism is important for vital processes such as vision, embryonic development, cell differentiation, and membrane and skin protection. The protein encoded by this gene is a key enzyme in beta-carotene metabolism to vitamin A. It catalyzes the oxidative cleavage of beta,beta-carotene into two retinal molecules.

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: BCO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR