Anti-DNAJC11 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-207
PRSI55-207EA 620 EUR
PRSI55-207
Anti-DNAJC11 Rabbit Polyclonal Antibody
Antikörper
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: DNAJC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR