Anti-LCE1A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-492
PRSI55-492EA 620 EUR
PRSI55-492
Anti-LCE1A Rabbit Polyclonal Antibody
Antikörper
LCE1A belongs to the late cornified envelope (LCE) gene cluster within the epidermal differentiation complex (EDC) on chromosome 1. The LCE cluster contains multiple conserved genes that encode stratum corneum proteins, and these genes are expressed relatively late during fetal assembly of the skin cornified envelope (Jackson et al., 2005 [PubMed 15854049]). For further information on the LCE gene cluster, see GENE FAMILY below.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: LCE1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR