Anti-DUS4L Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-573
PRSI55-573EA 620 EUR
PRSI55-573
Anti-DUS4L Rabbit Polyclonal Antibody
Antikörper
Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs (By similarity).

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: DUS4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR