Anti-BCO2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-590
PRSI55-590EA 620 EUR
PRSI55-590
Anti-BCO2 Rabbit Polyclonal Antibody
Antikörper
BCDO2 catalyzes the asymmetric oxidative cleavage of beta-carotene in carotene metabolism (Kiefer et al., 2001 [PubMed 11278918]).

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: BCO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR