Anti-HAUS3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-771
PRSI55-771EA 620 EUR
PRSI55-771
Anti-HAUS3 Rabbit Polyclonal Antibody
Antikörper
HAUS3 is 1 of 8 subunits of the 390-kD human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb 'augmentare,' meaning 'to increase.' The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 [PubMed 18443220]; Uehara et al., 2009 [PubMed 19369198]).

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.

Type: Primary
Antigen: HAUS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR