Anti-NCL Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-982
PRSI55-982EA 620 EUR
PRSI55-982
Anti-NCL Rabbit Polyclonal Antibody
Antikörper
Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For IF starting dilution is: 1:10~50.

Type: Primary
Antigen: NCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR