Anti-QTRTD1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-993
PRSI55-993EA 620 EUR
PRSI55-993
Anti-QTRTD1 Rabbit Polyclonal Antibody
Antikörper
Interacts with QTRT1 to form an active queuine tRNA-ribosyltransferase. This enzyme exchanges queuine for the guanine at the wobble position of tRNAs with GU(N) anticodons (tRNA-Asp, -Asn, -His and -Tyr), thereby forming the hypermodified nucleoside queuosine (Q) (7-(((4,5-cis-dihydroxy-2-cyclopenten-1-yl)amino)methyl)-7-deazaguanosine) (By similarity).

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: QTRTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR