Anti-VWA2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-127
PRSI56-127EA 620 EUR
PRSI56-127
Anti-VWA2 Rabbit Polyclonal Antibody
Antikörper
VWA2, also named as AMACO and CCSP2, is a novel candidate serological marker of colon neoplasia. VWA2 is found in the media of variesy of established cell lines of both fibroblast and epithelial origin.

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: VWA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR