Anti-PLCL2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-169
PRSI56-169EA 620 EUR
PRSI56-169
Anti-PLCL2 Rabbit Polyclonal Antibody
Antikörper
PLCL2 may play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum (By similarity).

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: PLCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR