Anti-B3GAT3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-189
PRSI56-189EA 620 EUR
PRSI56-189
Anti-B3GAT3 Rabbit Polyclonal Antibody
Antikörper
The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans.

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: B3GAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR