Anti-PCCA Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-351
PRSI56-351EA 620 EUR
PRSI56-351
Anti-PCCA Rabbit Polyclonal Antibody
Antikörper
The protein encoded by this gene is the alpha subunit of the heterodimeric mitochondrial enzyme Propionyl-CoA carboxylase. PCCA encodes the biotin-binding region of this enzyme. Mutations in either PCCA or PCCB (encoding the beta subunit) lead to an enzyme deficiency resulting in propionic acidemia. Multiple transcript variants encoding different isoforms have been found for this gene.

For IHC-P starting dilution is: 1:25 For WB starting dilution is: 1:2000.

Type: Primary
Antigen: PCCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR