Anti-DUS1L Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-478
PRSI56-478EA 620 EUR
PRSI56-478
Anti-DUS1L Rabbit Polyclonal Antibody
Antikörper
DUS1L catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs (By similarity).

  • For IHC-P starting dilution is: 1:10~50
  • For WB starting dilution is: 1:1000

Type: Primary
Antigen: DUS1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR