Anti-SLC9A2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-594
PRSI56-594EA 620 EUR
PRSI56-594
Anti-SLC9A2 Rabbit Polyclonal Antibody
Antikörper
SLC9A2 is involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Seems to play an important role in colonic sodium absorption.

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50

Type: Primary
Antigen: SLC9A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR