Anti-SNX4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-693
PRSI56-693EA 620 EUR
PRSI56-693
Anti-SNX4 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associated with the long isoform of the leptin receptor and with receptor tyrosine kinases for platelet-derived growth factor, insulin, and epidermal growth factor in cell cultures, but its function is unknown. This protein may form oligomeric complexes with family members. [provided by RefSeq].

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50

Type: Primary
Antigen: SNX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR