Anti-POFUT2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-696
PRSI56-696EA 620 EUR
PRSI56-696
Anti-POFUT2 Rabbit Polyclonal Antibody
Antikörper
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor (EGF; MIM 131530)-like repeats or thrombospondin (THBS; see MIM 188060) type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates (Luo et al., 2006 [PubMed 16464857]).

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50

Type: Primary
Antigen: POFUT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR