Anti-TAS2R1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-775
PRSI56-775EA 620 EUR
PRSI56-775
Anti-TAS2R1 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a member of a family of candidate taste receptors that are members of the G protein-coupled receptor superfamily and that are specifically expressed by taste receptor cells of the tongue and palate epithelia. This intronless taste receptor gene encodes a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is mapped to chromosome 5p15, the location of a genetic locus (PROP) that controls the detection of the bitter compound 6-n-propyl-2-thiouracil.

  • For WB starting dilution is: 1:1000

Type: Primary
Antigen: TAS2R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR