Anti-SSR2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-899
PRSI56-899EA 620 EUR
PRSI56-899
Anti-SSR2 Rabbit Polyclonal Antibody
Antikörper
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23. [provided by RefSeq].

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50
  • For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: SSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR