Anti-ACSS2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-934
PRSI56-934EA 620 EUR
PRSI56-934
Anti-ACSS2 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. The protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. Alternative splicing results in multiple transcript variants. [provided by RefSeq].

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50

Type: Primary
Antigen: ACSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR