Anti-PI15 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
56-994
PRSI56-994EA 620 EUR
PRSI56-994
Anti-PI15 Rabbit Polyclonal Antibody
Antikörper
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].

  • For WB starting dilution is: 1:1000
  • For IHC-P starting dilution is: 1:10~50
  • For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: PI15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR