Anti-RAMP3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
58-214
PRSI58-214EA 620 EUR
PRSI58-214
Anti-RAMP3 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 90-118
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR