Anti-HNRNPD Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
58-256
PRSI58-256EA 620 EUR
PRSI58-256
Anti-HNRNPD Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.

Type: Primary
Antigen: HNRNPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 57-85
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR