Anti-ITGB7 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
58-258
PRSI58-258EA 620 EUR
PRSI58-258
Anti-ITGB7 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: ITGB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 737-765
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR