Anti-FOXO4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
5817
PRSI5817EA 435 EUR
PRSI5817
Anti-FOXO4 Rabbit Polyclonal Antibody
Antikörper
FOXO4 Antibody: FOXO4 is a ubiquitously expressed protein member of a subfamily of the forkhead homeotic gene family of transcription factors and shuttles between the cytoplasm and nucleus. FOXO transcription factors are key players of cell fate decisions, metabolism, stress resistance, tumor suppression and are regulated by growth factors, oxidative stress or nutrient deprivation. In the absence of PI3K/AKT activation, FOXO4 localizes in the nucleus where it functions as a transcription factor. FOXO4 can also be phosphorylated by JNK following induction of reactive oxygen species (ROS), resulting in transcriptional activation and the induction of a negative feedback mechanism to counteract the ROS. It is through this mechanism that FOXO4 is thought to sensitize cancer cells to doxorubicin-mediated toxicity.

Type:
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR