Anti-OR2L8 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
59-346
PRSI59-346EA 620 EUR
PRSI59-346
Anti-OR2L8 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: OR2L8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 207-235
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR