Anti-MTF2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
59-772
PRSI59-772EA 620 EUR
PRSI59-772
Anti-MTF2 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: MTF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 289-317
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR