Anti-Vcl Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
61-325
PRSI61-325EA 620 EUR
PRSI61-325
Anti-Vcl Rabbit Polyclonal Antibody
Antikörper
For FACS starting dilution is: 1:25. For WB starting dilution is: 1:2000.

Type: Primary
Antigen: Vcl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR