Anti-TEC Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-168
PRSI63-168EA 620 EUR
PRSI63-168
Anti-TEC Rabbit Polyclonal Antibody
Antikörper
For IF starting dilution is: 1:10~50. For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: TEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 175-205
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR