Anti-GRK7 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-174
PRSI63-174EA 620 EUR
PRSI63-174
Anti-GRK7 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: GRK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 508-538
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR